1-Bromo-4-(difluoromethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004836-1G 004836-5G 004836-25G
FLUO004836-1GEA 0 GBP
FLUO004836-1G FLUO004836-5G FLUO004836-25G
1-Bromo-4-(difluoromethoxy)benzene
1-Bromo-4-(difluoromethoxy)benzene
Formula: C₇H₅BrF₂O
MDL Number: MFCD00221465
CAS Number: 5905-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR