2-Bromo-4-fluoroanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002823-100G
002823-25G
FLUO002823-100GEA
0
GBP
FLUO002823-100G
FLUO002823-25G
2-Bromo-4-fluoroanisole
2-Bromo-4-fluoroanisole
Formula:
C₇H₆BrFO Boiling Pt: 274 °C (1013 hPa) Melting Pt: –27 °C Density: 1,58 g/cm³ (20 °C) |
MDL Number:
MFCD00012014 CAS Number: 452-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...