4-Bromo-2-fluoroanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004207-100G
004207-250G
FLUO004207-100GEA
0
GBP
FLUO004207-100G
FLUO004207-250G
4-Bromo-2-fluoroanisole
4-Bromo-2-fluoroanisole
Formula:
C₇H₆BrFO Boiling Pt: 215 °C (1013 hPa) Melting Pt: 16 °C Density: 1,59 g/cm³ (20 °C) |
MDL Number:
MFCD00011710 CAS Number: 2357-52-0 EINECS: 219-096-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...