4-Bromo-2-fluoroanisole ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-bromo-2-fluoro-1-methoxybenzene
A10948.14
A10948.22
A10948.14EA
68.8
GBP
A10948.14
A10948.22
4-Bromo-2-fluoroanisole ≥97%
4-Bromo-2-fluoroanisole
Purity: 97%
CAS No.: 2357-52-0
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Flash Point: 98°(208°F)
Refractive Index: 1.5450
Boiling Point: 208°
Beilstein Reference: 3240714
MDL No.: MFCD00011710
Formula:
C₇H₆BrFO MW: 205.03 g/mol Boiling Pt: 208 °C Melting Pt: 16 Density: 1.590 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011710 CAS Number: 2357-52-0 EINECS: 219-096-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...