4-Bromo-3,5-difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF348.5 CFF348.25 017383-5G 017383-25G 017383-1G 017383-100G
CHMPCFF348.5EA 0 GBP
CHMPCFF348.5 CHMPCFF348.25 FLUO017383-5G FLUO017383-25G FLUO017383-1G FLUO017383-100G
4-Bromo-3,5-difluoroaniline
4-Bromo-3,5-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
MDL Number: MFCD00236593
CAS Number: 203302-95-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR