4-Bromo-2-fluoroaniline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
304220-10G
304220-50G
SIAL304220-10GEA
70.9
GBP
SIAL304220-10G
SIAL304220-50G
4-Bromo-2-fluoroaniline, Sigma-Aldrich®
4-Bromo-2-fluoroaniline
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 75…80 °C (0.3 mmHg) Melting Pt: 41…42 °C |
MDL Number:
MFCD00010221 CAS Number: 367-24-8 EINECS: 206-685-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...