2-Bromo-4-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003144-25G 003144-100G
FLUO003144-25GEA 0 GBP
FLUO003144-25G FLUO003144-100G
2-Bromo-4-fluoroaniline
2-Bromo-4-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 46…47 °C (0.05 mmHg)
Melting Pt: 41
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00042462
CAS Number: 1003-98-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR