2-Bromo-4,6-difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: BDFA

001702-25G 001702-100G 001702.500G
FLUO001702-25GEA 0 GBP
FLUO001702-25G FLUO001702-100G FLUO001702.500G
2-Bromo-4,6-difluoroaniline
2-Bromo-4,6-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 42…42.5 °C
MDL Number: MFCD00009639
CAS Number: 444-14-4
EINECS: 429-430-0
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR