1-Bromo-4-fluoro-2-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013232-25G 013232-5G
FLUO013232-25GEA 0 GBP
FLUO013232-25G FLUO013232-5G
1-Bromo-4-fluoro-2-iodobenzene
1-Bromo-4-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
MDL Number: MFCD00042578
CAS Number: 202865-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR