1-Bromo-3-fluoro-5-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Fluoro-5-iodobromobenzene

023889-1G 023889-5G
FLUO023889-1GEA 0 GBP
FLUO023889-1G FLUO023889-5G
1-Bromo-3-fluoro-5-iodobenzene
1-Bromo-3-fluoro-5-iodobenzene
Formula: C₆H₃BrFI
MDL Number: MFCD06657969
CAS Number: 845866-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR