3-Bromo-5-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF843.1 FL5475.500 005475-5G 005475-1G
CHMPCFF843.1EA 0 GBP
CHMPCFF843.1 CHMPFL5475.500 FLUO005475-5G FLUO005475-1G
3-Bromo-5-fluorobenzoic acid
3-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD01569540
CAS Number: 176548-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR