2-Bromo-6-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF840.1 005472-1G 005472-500MG 005472-5G
CHMPCFF840.1EA 0 GBP
CHMPCFF840.1 FLUO005472-1G FLUO005472-500MG FLUO005472-5G
2-Bromo-6-fluorobenzoic acid
2-Bromo-6-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MDL Number: MFCD01569539
CAS Number: 2252-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR