1-Bromo-2,3,4-trifluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003864-25G 003864-100G
FLUO003864-25GEA 0 GBP
FLUO003864-25G FLUO003864-100G
1-Bromo-2,3,4-trifluorobenzene
1-Bromo-2,3,4-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
MDL Number: MFCD00013248
CAS Number: 176317-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR