1,3-Dibromotetrafluorobenzene ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,3-Dibromo-2,4,5,6-tetrafluorobenzene
A17392.06EA
138
GBP
A17392.06
A17392.03
A17392.14
1,3-Dibromotetrafluorobenzene ≥98%
1,3-Dibromotetrafluorobenzene
Purity: 98+%
CAS No.: 1559-87-1
Molecular Formula: C6Br2F4
Formula Weight: 307.87
Flash Point: 92°(198°F)
Refractive Index: 1.5160
Boiling Point: 85-88°/18mm
MDL No.: MFCD00031533
Formula:
C₆Br₂F₄ MW: 307.87 g/mol Boiling Pt: 85…88 °C (18 mmHg) Density: 2.180 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00031533 CAS Number: 1559-87-1 EINECS: 216-326-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...