4-Bromobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: p-Bromobenzoyl chloride

013058-100G 013058-25G
FLUO013058-100GEA 0 GBP
FLUO013058-100G FLUO013058-25G
4-Bromobenzoyl chloride
4-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 36 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000683
CAS Number: 586-75-4
EINECS: 209-580-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR