2-Bromobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride

001051-25G 001051-100G
FLUO001051-25GEA 0 GBP
FLUO001051-25G FLUO001051-100G
2-Bromobenzoyl chloride
2-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 11 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000655
CAS Number: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR