1-Bromo-2,5-dichloro-3-fluorobenzene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L16713.03 L16713.14
L16713.03EA 15.5 GBP
L16713.03 L16713.14
1-Bromo-2,5-dichloro-3-fluorobenzene ≥97%
1-Bromo-2,5-dichloro-3-fluorobenzene

Purity: 97% CAS No.: 202865-57-4 Molecular Formula: C6H2BrCl2F Formula Weight: 243.89 Refractive Index: 1.5757

Formula: C₆H₂BrCl₂F
Storage Temperature: Ambient
MDL Number: MFCD00142585
CAS Number: 202865-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR