2-Bromo-6-chlorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019332-1G 019332-5G
FLUO019332-1GEA 0 GBP
FLUO019332-1G FLUO019332-5G
2-Bromo-6-chlorobenzoic acid
2-Bromo-6-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00672929
CAS Number: 93224-85-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR