2-Bromo-1,3,5-trichlorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-2,4,6-trichlorobenzene

018322-5G 018322-25G
FLUO018322-5GEA 0 GBP
FLUO018322-5G FLUO018322-25G
2-Bromo-1,3,5-trichlorobenzene
2-Bromo-1,3,5-trichlorobenzene
Formula: C₆H₂BrCl₃
MW: 260.34 g/mol
MDL Number: MFCD00060653
CAS Number: 19393-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR