2,5-Dimethoxyphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010990-1G 010990-5G 010990-25G
FLUO010990-1GEA 0 GBP
FLUO010990-1G FLUO010990-5G FLUO010990-25G
2,5-Dimethoxyphenylboronic acid
2,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MDL Number: MFCD01318181
CAS Number: 107099-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR