2,4-Dimethoxyphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010989-1G 010989-5G
FLUO010989-1GEA 0 GBP
FLUO010989-1G FLUO010989-5G
2,4-Dimethoxyphenylboronic acid
2,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MDL Number: MFCD01074590
CAS Number: 133730-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR