3-Cyanophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

FL10787.2 010787-5G 010787-25G
CHMPFL10787.2EA 0 GBP
CHMPFL10787.2 FLUO010787-5G FLUO010787-25G
3-Cyanophenylboronic acid
3-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MDL Number: MFCD01318967
CAS Number: 150255-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR