Phenylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
010816-50G
010816-100G
FLUO010816-50GEA
0
GBP
FLUO010816-50G
FLUO010816-100G
Phenylboronic acid
Phenylboronic acid
Formula:
C₆H₇BO₂ Melting Pt: 217…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002103 CAS Number: 98-80-6 EINECS: 202-701-9 Merck Index: 13,01068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...