2,2-Difluoro-1,3-benzodioxole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole

007845-10G 007845-25G 007845-5G
FLUO007845-10GEA 0 GBP
FLUO007845-10G FLUO007845-25G FLUO007845-5G
2,2-Difluoro-1,3-benzodioxole
2,2-Difluoro-1,3-benzodioxole
Formula: C₇H₄F₂O₂
Boiling Pt: 129…130 °C (1013 hPa)
Density: 1.3079 g/cm³ (20 °C)
MDL Number: MFCD00236217
CAS Number: 1583-59-1
EINECS: 216-431-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR