5-Fluoroisatin

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Fluoroindoline-2,3-dione, 5-Fluoro-1H-indole-2,3-dione

CFF622.2.5 008155-5G 008155-25G
CHMPCFF622.2.5EA 0 GBP
CHMPCFF622.2.5 FLUO008155-5G FLUO008155-25G
5-Fluoroisatin
5-Fluoroisatin
Formula: C₈H₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR