2-Methylpyrrolidine ≥97%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Purity: 97%
CAS No.: 765-38-8
Molecular Formula: C5H11N
Formula Weight: 85.15
Flash Point: 2°(37°F)
Physical Form: Liquid
Density: 0.834
Refractive Index: 1.4370
Odour: Amine-like
UN No.: 2733
MDL No.: MFCD00014491
Formula:
C₅H₁₁N Density: 0.834 Flash Pt: 2 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014491 CAS Number: 765-38-8 EINECS: 212-144-3 UN: 2733 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...