4-Hydroxy-1-methylpiperidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
023287-100G
023287-25G
FLUO023287-100GEA
0
GBP
FLUO023287-100G
FLUO023287-25G
4-Hydroxy-1-methylpiperidine
4-Hydroxy-1-methylpiperidine
Formula:
C₆H₁₃NO Boiling Pt: 200 °C (1013 hPa) Melting Pt: 29…31 °C Density: 0,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006500 CAS Number: 106-52-5 EINECS: 203-406-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...