Pyrrolidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Tetrahydropyrrole

M-Clarity™ quality level = MQ200

394238-100ML 394238-1L
SIAL394238-100MLEA 73.6 GBP
SIAL394238-100ML SIAL394238-1L
Pyrrolidine, Sigma-Aldrich®
Pyrrolidine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 87
Melting Pt: –63 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 3
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3 II
Merck Index: 12,08199

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR