N-Hydroxysuccinimide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

130672-100G 130672-250G 130672-25G 130672-5G
SIAL130672-100GEA 103 GBP
SIAL130672-100G SIAL130672-250G SIAL130672-25G SIAL130672-5G
N-Hydroxysuccinimide, Sigma-Aldrich®
N-Hydroxysuccinimide
Formula: C₄H₅NO₃
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 93…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005516
CAS Number: 6066-82-6
EINECS: 228-001-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR