3-Cyclohexylpropyne
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009112-5G
009112-25G
FLUO009112-5GEA
0
GBP
FLUO009112-5G
FLUO009112-25G
3-Cyclohexylpropyne
3-Cyclohexylpropyne
Formula:
C₉H₁₄ Boiling Pt: 48 °C (1013 hPa) Density: 0.844 g/cm³ (18 °C) |
MDL Number:
MFCD00041562 CAS Number: 17715-00-3 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...