3-Butyne-1-ol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

009077-10G 009077-50G
FLUO009077-10GEA 0 GBP
FLUO009077-10G FLUO009077-50G
3-Butyne-1-ol
3-Butyne-1-ol
Formula: C₄H₆O
Boiling Pt: 128.8 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.9262 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR