2-Butyne-1-ol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

009076-10G 009076-50G 009076-25G
FLUO009076-10GEA 0 GBP
FLUO009076-10G FLUO009076-50G FLUO009076-25G
2-Butyne-1-ol
2-Butyne-1-ol
Formula: C₄H₆O
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR