5-Hexynoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

544000-25G 544000-5G
SIAL544000-25GEA 449 GBP
SIAL544000-25G SIAL544000-5G
5-Hexynoic acid, Sigma-Aldrich®
5-Hexynoic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 41 °C
Density: 1.0372 g/cm³ (20 °C)
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR