4-Pentynoic acid 95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

213730010 213730050
ACRO213730010EA 34.9 GBP
ACRO213730010 ACRO213730050
4-Pentynoic acid 95%
4-Pentynoic acid
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 110 °C (30 mmHg)
Melting Pt: 54…58 °C
Flash Pt: 75 °C
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 54 to 60 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR