2-Butynoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
303666-1G
303666-5G
SIAL303666-1GEA
38.8
GBP
SIAL303666-1G
SIAL303666-5G
2-Butynoic acid, Sigma-Aldrich®
2-Butynoic acid
Formula:
C₄H₄O₂ MW: 84.07 g/mol Boiling Pt: 203 Melting Pt: 78 Density: 0.9641 g/cm³ (20 °C) |
MDL Number:
MFCD00004363 CAS Number: 590-93-2 EINECS: 209-695-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...