2-Butynoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
009075-5G
009075-25G
FLUO009075-5GEA
0
GBP
FLUO009075-5G
FLUO009075-25G
2-Butynoic acid
2-Butynoic acid
Formula:
C₄H₄O₂ MW: 84.07 g/mol Boiling Pt: 203 Melting Pt: 78 Density: 0.9641 g/cm³ (20 °C) |
MDL Number:
MFCD00004363 CAS Number: 590-93-2 EINECS: 209-695-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...