3-Butyne-2-one
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
But-3-yn-2-one
009078-5G
009078-25G
FLUO009078-5GEA
0
GBP
FLUO009078-5G
FLUO009078-25G
3-Butyne-2-one
3-Butyne-2-one
Formula:
C₄H₄O Boiling Pt: 85 °C (1013 hPa) Density: 0.886 g/cm³ (15 °C) |
MDL Number:
MFCD00008775 CAS Number: 1423-60-5 EINECS: 215-834-2 UN: 1992 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...