5-Chloro-1-pentyne, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
244376-25G
244376-5G
SIAL244376-25GEA
163
GBP
SIAL244376-25G
SIAL244376-5G
5-Chloro-1-pentyne, Sigma-Aldrich®
5-Chloro-1-pentyne
Formula:
C₅H₇Cl MW: 102.56 g/mol Boiling Pt: 67 °C (1013 hPa) Density: 0.96 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00001014 CAS Number: 14267-92-6 EINECS: 238-159-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...