1-Bromo-2-butyne

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009057-5G 009057-25G
FLUO009057-5GEA 0 GBP
FLUO009057-5G FLUO009057-25G
1-Bromo-2-butyne
1-Bromo-2-butyne
Formula: C₄H₅Br
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR