4-Fluorocinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2E)-3-(4-fluorophenyl)prop-2-enoic acid, p-Fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, trans-4-Fluorocinnamic acid

001180-25G 001180-100G 001180-250G
FLUO001180-25GEA 0 GBP
FLUO001180-25G FLUO001180-100G FLUO001180-250G
4-Fluorocinnamic acid
4-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 209 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00004395
CAS Number: 459-32-5
EINECS: 207-288-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR