trans-2,4-Difluorocinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (E)-3-(2,4-Difluorophenyl)acrylic acid, 2,4-Difluorocinnamic acid

004805-1G 004805-5G 004805-25G
FLUO004805-1GEA 0 GBP
FLUO004805-1G FLUO004805-5G FLUO004805-25G
trans-2,4-Difluorocinnamic acid
trans-2,4-Difluorocinnamic acid
Formula: C₉H₆F₂O₂
MDL Number: MFCD00010317
CAS Number: 94977-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR