trans-2,3-Difluorocinnamic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(E)-3-(2,3-Difluorophenyl)acrylic acid
004804-1G
004804-5G
004804-10G
FLUO004804-1GEA
0
GBP
FLUO004804-1G
FLUO004804-5G
FLUO004804-10G
trans-2,3-Difluorocinnamic acid
trans-2,3-Difluorocinnamic acid
Formula:
C₉H₆F₂O₂ |
MDL Number:
MFCD00061291 CAS Number: 207981-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...