Cinnamic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

W228826-100G-K W228826-1KG-K
SIALW228826-100G-KEA 162 GBP
SIALW228826-100G-K SIALW228826-1KG-K
Cinnamic acid, Sigma-Aldrich®
Cinnamic acid
Formula: C₉H₈O₂
MW: 148.15 g/mol
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR