Cinnamic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ300
W228818-10KG-K
W228818-1KG-K
W228818-25KG-K
SIALW228818-10KG-KEA
636
GBP
SIALW228818-10KG-K
SIALW228818-1KG-K
SIALW228818-25KG-K
Cinnamic acid, Sigma-Aldrich®
Formula:
C₉H₈O₂ |
MDL Number:
MFCD00004369 CAS Number: 140-10-3 EINECS: 205-398-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...