trans-Cinnamic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid
M-Clarity™ quality level = MQ200
133760-100G
133760-2KG
133760-500G
SIAL133760-100GEA
57
GBP
SIAL133760-100G
SIAL133760-2KG
SIAL133760-500G
trans-Cinnamic acid, Sigma-Aldrich®
trans-Cinnamic acid
Formula:
C₉H₈O₂ Boiling Pt: 300 °C (1013 hPa) Melting Pt: 131…134 °C Density: 1.25 g/cm³ (4 °C) |
MDL Number:
MFCD00004369 CAS Number: 140-10-3 EINECS: 205-398-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...