2-Bromo-4-fluorocinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013229-1G 013229-5G
FLUO013229-1GEA 0 GBP
FLUO013229-1G FLUO013229-5G
2-Bromo-4-fluorocinnamic acid
2-Bromo-4-fluorocinnamic acid
Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
CAS Number: 289038-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR