4-Isopropylcinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(4-Isopropylphenyl)acrylic acid

011177-25G 011177-100G
FLUO011177-25GEA 0 GBP
FLUO011177-25G FLUO011177-100G
4-Isopropylcinnamic acid
4-Isopropylcinnamic acid
Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00016544
CAS Number: 3368-21-6
EINECS: 222-138-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR