1-Chloro-3-methyl-2-butene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
303259-25G
303259-5G
SIAL303259-25GEA
308
GBP
SIAL303259-25G
SIAL303259-5G
1-Chloro-3-methyl-2-butene, Sigma-Aldrich®
1-Chloro-3-methyl-2-butene
Formula:
C₅H₉Cl Boiling Pt: 25 °C (1013 hPa) Density: 0.9273 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000982 CAS Number: 503-60-6 EINECS: 207-972-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...