trans-3-Hexen-1-ol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
trans-Hex-3-en-1-ol
M-Clarity™ quality level = MQ100
224715-25G
224715-5G
SIAL224715-25GEA
305
GBP
SIAL224715-25G
SIAL224715-5G
trans-3-Hexen-1-ol, Sigma-Aldrich®
trans-3-Hexen-1-ol
Formula:
C₆H₁₂O Boiling Pt: 155…156 °C (1013 hPa) Melting Pt: –60 °C Density: 1.515 g/cm³ (20 °C) |
MDL Number:
MFCD00002960 CAS Number: 928-97-2 EINECS: 213-193-3 UN: 1987 ADR: 3,III Merck Index: 12,04737 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...