3-(1,1,2,2-Tetrafluoroethoxy)propene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010391-25G 010391-1G 010391-5G
FLUO010391-25GEA 0 GBP
FLUO010391-25G FLUO010391-1G FLUO010391-5G
3-(1,1,2,2-Tetrafluoroethoxy)propene
3-(1,1,2,2-Tetrafluoroethoxy)propene
Formula: C₅H₆F₄O
MW: 158.1 g/mol
Boiling Pt: 74 °C (739 mmHg)
Density: 1.204 g/cm³ (20 °C)
MDL Number: MFCD00043822
CAS Number: 1428-33-7
EINECS: 215-847-3
UN: 1992

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR